Skip to Content
MilliporeSigma
All Photos(1)

Documents

C112040

Sigma-Aldrich

DMT-dC(tac) Phosphoramidite

configured for MerMade

Synonym(s):

DMT-dC(tac) amidite, N-[2-[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H62N5O9P
CAS Number:
Molecular Weight:
920.04
UNSPSC Code:
41116105
NACRES:
NA.51

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

compatibility

configured for MerMade

nucleoside profile

base: deoxycytidine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast

storage temp.

-10 to -25°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(c2ccccc2)(c3ccccc3)c4ccccc4)N5C=CC(NC(=O)COc6ccc(cc6)C(C)(C)C)=NC5=O

InChI

1S/C49H58N5O7P/c1-35(2)54(36(3)4)62(59-31-17-29-50)61-42-32-46(53-30-28-44(52-47(53)56)51-45(55)34-57-41-26-24-37(25-27-41)48(5,6)7)60-43(42)33-58-49(38-18-11-8-12-19-38,39-20-13-9-14-21-39)40-22-15-10-16-23-40/h8-16,18-28,30,35-36,42-43,46H,17,31-34H2,1-7H3,(H,51,52,55,56)/t42-,43+,46+,62?/m0/s1

InChI key

NHKNJNORWFDRBB-ROEGVEHFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service