C1105
Z-Gly-Pro-Arg 4-methoxy-2-naphthylamide trifluoroacetate salt
≥97% (HPLC), powder
Synonym(s):
Z-GPR-MNA TFA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C34H40F3N7O8
Molecular Weight:
731.72
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
form
powder
color
white
solubility
DMF or absolute ethanol: 20 mM
storage temp.
−20°C
Amino Acid Sequence
Z-Gly-Pro-Arg-4M2NA
Application
Fluorogenic substrate for granzyme A.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
F Velotti et al.
European journal of immunology, 22(4), 1049-1053 (1992-04-01)
Granzymes A-G are a family of serine proteases localized in the cytoplasmic granules of cytotoxic T lymphocytes (Masson, D. et al. Cell 1987. 49: 679) and granzyme A is secreted by T lymphocytes in response to antigenic stimulation. Granzyme A
M J Smyth et al.
The Journal of biological chemistry, 267(34), 24418-24425 (1992-12-05)
We have purified a 30-kDa serine protease (designated RNK-Met-1) from the granules of the rat large granular lymphocyte leukemia cell line (RNK-16) that hydrolytically cleaves model peptide substrates after methionine, leucine, and norleucine (Met-ase activity). Utilizing molecular sieve chromatography, heparin-agarose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service