Skip to Content
MilliporeSigma
All Photos(1)

Documents

C0163

Sigma-Aldrich

Cholesteryl dodecanoate

≥98% (HPLC; detection at 205 nm)

Synonym(s):

3β-Hydroxy-5-cholestene 3-dodecanoate, 5-Cholesten-3β-ol 3-dodecanoate, Cholesteryl laurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68O2
CAS Number:
Molecular Weight:
568.96
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC; detection at 205 nm)

suitability

suitable for HPLC marker

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@]1([H])[C@@]2(C)CC[C@@]3([H])[C@@]([C@]2([H])CC1)([H])CC=C4C[C@@H](OC(CCCCCCCCCCC)=O)CC[C@@]43C

InChI

1S/C39H68O2/c1-7-8-9-10-11-12-13-14-15-19-37(40)41-32-24-26-38(5)31(28-32)20-21-33-35-23-22-34(30(4)18-16-17-29(2)3)39(35,6)27-25-36(33)38/h20,29-30,32-36H,7-19,21-28H2,1-6H3

InChI key

RMLFYKFCGMSLTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander D Dergunov et al.
Archives of biochemistry and biophysics, 564, 211-218 (2014-12-03)
The transfer of cholesteryl ester by recombinant cholesteryl ester transfer protein (CETP) between reconstituted discoidal high-density lipoprotein (rHDL) was studied. Particles contained apolipoprotein A-I, unsaturated POPC or saturated DPPC and cholesteryl ester as cholesteryl 1-pyrenedecanoate (CPD) or cholesteryl laurate (CL)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service