Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR781413

BRAND® PCR caps, strips of 8

clear, flat polypropylene

Synonym(s):

Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps

Sign Into View Organizational & Contract Pricing

Select a Size

300 EA
$257.00

$257.00


Please contact Customer Service for Availability


Select a Size

Change View
300 EA
$257.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

$257.00


Please contact Customer Service for Availability

material

flat polypropylene

feature

RNase and DNase free

packaging

pkg of 25 × 12 strips (300 total)

manufacturer/tradename

BRAND 781413

technique(s)

PCR: suitable

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

  • BRAND® PCR caps, strips of 8 are transparent, flat caps and skirted.
  • Applicable for sealing 96-well skirted PCR plates (781377-78) and qPCR applications.
  • Free of DNase, RNase and RNA.

Features and Benefits

  • Effective protection against evaporation and contamination
  • The right cap shape for your application
  • Easy to open and close without risk of contamination

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service