Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR701458

BRAND® Reagent Reservoirs for multichannel pipettes and automatic systems

1 column, low profile, column capacity 50 mL, flat bottom

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.01

material

colorless
flat bottom
polypropylene

sterility

non-sterile

feature

RNase and DNase free
1 column
low profile

packaging

pack of 10

manufacturer/tradename

BRAND 701458

capacity

50 mL

column capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

These reagent reservoirs were designed especially for use with automated liquid handling platforms. They can also be used with manual systems. Different partitions, bottom styles and profiles provide great application flexibility.

Features and Benefits

  • ANSI/SLAS standard format Compatible with nearly all liquid handling robots and other automated dispensing systems
  • 5 different types for use with multichannel pipettes and automatic systems
  • Excellent stability and chemical resistance made from high-quality polypropylene
  • Very low dead volume due to pyramid bottoms
  • Free from RNases and DNases
  • Autoclavable (121 °C, 20 min)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service