Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR26539

BRAND®, accu-jet® S wall support

White

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121514
NACRES:
NB.04

material

PBT
PC

packaging

pkg of 1 ea

manufacturer/tradename

BrandTech 26539

color

White

General description

Accessories for BRAND®, accu-jet® S pipette controller

Application

storage of Brand®, accu-jet® S pipette controller

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service