Key Documents
BMSIPD9600BD5
Sigma-Aldrich
Ball Dropper to Fill 96 BeadBlaster 96 Well Plate
diam. 5 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
acrylic
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPD9600-BD5
diam.
5 mm
General description
This ball dropper is used to easily fill 96 well plates with 1 bead per well.
Application
Homogenization/milling/grinding of samples
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)