Recommended Products
AC/DC input
110 V / 120 V, US 3-pin plug
material
stainless steel device (Coated Steel Ext Stainless Steel Int)
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
B Medical Systems
parameter
-20--41 °C temp. range
H × W × D
78.27 in. × 39.06 in. × 40.91 in.
capacity
26.38 ft3
weight
603.28 lb
shipped in
ambient
storage temp.
room temp
General description
- Premium - Upright, Laboratory Freeze
- 5 Wire Shelves
- 4 Casters with integrated feet
- 7" touchscreen Displaywith Proximity Card Access Control
- 2 Interior Doors
- Alarm System w/ Battery Back-Up
- USB Data Port
- Door Mounted Control Panel
- Electronic Door Access
Application
Laboratory Refrigeration
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service