Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0027

Sigma-Aldrich

BMS-870145

≥98% (HPLC)

Synonym(s):

BMS 870145, BMS-870145-01, N-[2-[4-(4-Chlorophenyl)-5-fluoro-2,3-dihydro-7-hydroxy-3,3-dimethyl-1H-indol-1-yl]phenyl]-N′-[4-(trifluoromethoxy)phenyl]-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24ClF4N3O3
CAS Number:
Molecular Weight:
585.98
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

FC1=CC(O)=C2C(C(C)(C)CN2C3=C(NC(NC4=CC=C(OC(F)(F)F)C=C4)=O)C=CC=C3)=C1C5=CC=C(Cl)C=C5

Biochem/physiol Actions

BMS-870145 is an orally available, potent and selective P2Y1 purinergic receptor antagonist.
BMS-870145 is an orally available, potent and selective P2Y1 purinergic receptor antagonist. BMS-870145 demonstrated similar antithrombotic efficacy with less bleeding compared with P2Y12 antagonist prasugrel in rabbits.

Features and Benefits

BMS-870145 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service