Skip to Content
MilliporeSigma
All Photos(1)

Documents

BM0023

Sigma-Aldrich

BMS-770767

≥98% (HPLC)

Synonym(s):

4-[8-(2-Chlorophenoxy)-1,2,4-triazolo[4,3-a]pyridin-3-yl]-bicyclo[2.2.1]heptan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18ClN3O2
CAS Number:
Molecular Weight:
355.82
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

ClC1=C(OC2=CC=CN3C2=NN=C3[C@]4(C5)CC[C@]5(O)CC4)C=CC=C1

InChI

1S/C19H18ClN3O2/c20-13-4-1-2-5-14(13)25-15-6-3-11-23-16(15)21-22-17(23)18-7-9-19(24,12-18)10-8-18/h1-6,11,24H,7-10,12H2/t18-,19+

InChI key

HNXGIFYHJKEXNA-KDURUIRLSA-N

Biochem/physiol Actions

BMS-770767 is an inhibitor of 11-betahydroxysteroid dehydrogenase type I (11β-HSD1), which is a regulator of glucocorticoid activity and thought to play a possible role in metabolic syndrome. 11β-HSD1 can convert cortisol to the inactive metabolite cortisone, or catalyze the reverse reaction, converting inactive cortisone into active cortisol in cells. BMS-770767 was in clinical trials for hypercholesterolemia and Type 2 diabetes.

Features and Benefits

BMS-770767 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service