Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0021

Sigma-Aldrich

BMY-14802

≥97% (HPLC)

Synonym(s):

α-(4-Fluorophenyl)-4-(5-fluoro-2-pyrimidinyl)-1-piperazine butanol, (±)-BMY 14802, BMS 181100, BMY-014802, BMY14802

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22F2N4O
CAS Number:
Molecular Weight:
348.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

FC1=CC=C(C(O)CCCN2CCN(C3=NC=C(F)C=N3)CC2)C=C1

InChI

1S/C18H22F2N4O/c19-15-5-3-14(4-6-15)17(25)2-1-7-23-8-10-24(11-9-23)18-21-12-16(20)13-22-18/h3-6,12-13,17,25H,1-2,7-11H2

InChI key

ZXUYYZPJUGQHLQ-UHFFFAOYSA-N

Biochem/physiol Actions

BMY-14802 is a potent and selective sigma-1 (σ1) receptor antagonist with an IC50 value of approximately 100 nM. BMY-14802 shows low affinity at serotonin (5-HT) 1A and adrenergic σ1 receptors and minimal activity at dopamine receptors. BMY-14802 has exhibited antipsychotic activity in rodent models and anti-dyskinetic effects against L-DOPA with preservation of antiparkinsonian efficacy.

Features and Benefits

BMY-14802 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service