Skip to Content
MilliporeSigma
All Photos(1)

Documents

BM0012

Sigma-Aldrich

BMS-191095 hydrochloride

≥98% (HPLC)

Synonym(s):

(3R,4S)-4-[(4-Chlorophenyl)(1H-imidazol-2-ylmethyl)amino]-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-6-carbonitrile monohydrochloride, (3R-trans)-4-[(4-Chlorophenyl)(1H-imidazol-2-ylmethyl)amino]-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-Benzopyran-6-carbonitrile monohydrochloride, BMS 191095 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21ClN4O2 · HCl
CAS Number:
Molecular Weight:
445.34
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC1(C)OC2=CC=C(C#N)C=C2[C@H](N(CC3=NC=CN3)C4=CC=C(Cl)C=C4)[C@H]1O.[H]Cl

InChI

1S/C22H21ClN4O2.ClH/c1-22(2)21(28)20(17-11-14(12-24)3-8-18(17)29-22)27(13-19-25-9-10-26-19)16-6-4-15(23)5-7-16;/h3-11,20-21,28H,13H2,1-2H3,(H,25,26);1H/t20-,21+;/m0./s1

InChI key

TZLMTXNZECPICU-JUDYQFGCSA-N

Biochem/physiol Actions

BMS-191095 is a potent and selective mitochondrial ATP-sensitive K1 channel (KATP) channel opener devoid of peripheral vasodilating activity. BMS-191095 improves postischemic recovery of function and reduced lactate dehydrogenase release in the isolated rat hearts.

Features and Benefits

BMS-191095 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service