Skip to Content
Merck
All Photos(2)

Key Documents

BCS-170

CoolCell FTS30

Freezing container, for 30 x 1 mL or 2 mL cryogenic vials, purple

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41103025

feature

capacity 30  vial(s)
for vials 1 ‑ 2 mL
for 30 x 1 mL or 2 mL cryogenic vials

manufacturer/tradename

BioCision BCS-170

diam. × H

16.5 cm × 11.5 cm

well diam.

12.3 mm

color

purple

Looking for similar products? Visit Product Comparison Guide

General description

CoolCell FTS30 alcohol-free cell freezing container provides a highly reproducible -1°C/minute freezing rate when used in a -80°C freezer. No alcohol or any other fluids required. Unique patent-pending micro-convection design takes in cold freezer air through a bottom intake port, evenly distributes the cold air around all vials, then releases warm air through the top port. The insulative outer housing and thermo-conductive solid state core ensure consistent heat removal to all vials throughout the freezing period. CoolCell containers are unbreakable, open easily when frozen, and are not cold to the touch when frozen - no "frosty" fingers. Proven for use with stem cells, primary cells and cell lines.

Note: This product is NOT STACKABLE.

Save money and go green!
CoolCell containers replace isopropanol-based freezing containers which can consume up to 12L per year of IPA and cost hundred of dollars per unit per year to maintain. CoolCell containers instantly save money and eliminate hazardous solvents from the cell cryopreservation process. Calculate your cost savings here.

Holds 30 1.0 mL or 2.0 mL cryogenic vials. Cryogenic vials fill volume: 1.0 mL per vial. Fill empty wells with CoolCell Filler Vials.

Legal Information

CoolCell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service