Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAF370150000

CultureTemp® Warming Plate

AC/DC input 120 V AC, US 2-pin plug

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$769.00

$769.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$769.00

About This Item

UNSPSC Code:
41102406

$769.00


Please contact Customer Service for Availability

material

anodized aluminum plate

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37015-0000

depth × W × H

25.4 × 24.9 × 5.7 cm

plate depth × W

17.8 × 20.3 cm

Looking for similar products? Visit Product Comparison Guide

General description

Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells at an optimal 37°C (+/-1°C ) outside of the incubator. The unit is water resistant and safe for cleaning with Ethanol or IPA.

Features and Benefits

  • Keep dishes outside the incubator longer; 37°C plate protects cell viability; no need to return to the incubator repeatedly
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Temperature remains uniform across the entire anodized aluminum warming plate t
  • Small footprint fits easily in a cell culture hood or on the workbench
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • 15-minute count down timer keeps track of time sensitive applications

Legal Information

CultureTemp is a registered trademark of SP Scienceware, Division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service