Key Documents
BAF198070000
Bel-Art® Centrifuge Tubes, Rimless
tube capacity × L 45 mL × 104 mm, polypropylene, pack of 12 tubes
Synonym(s):
round bottom tube
About This Item
UNSPSC Code:
41121703
NACRES:
NB.22
material
polypropylene
round bottom
feature
autoclavable
packaging
pack of 12 tubes
manufacturer/tradename
Bel-Art F19807-0000
tube capacity × L
45 mL × 104 mm
General description
Rimless polypropylene centrifuge tubes for use in centrifuges up to 50,000 xg without refrigeration. Autoclavable at 121ºC (250ºF).
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)