Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAF135570000

Charcoal Disks

for 100 mm Petri Dish, pack of 50 discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123000
NACRES:
NB.14

form

disc

packaging

pack of 50 discs

manufacturer/tradename

Bel-Art F13557-0000

General description

Activated charcoal impregnated into each Scienceware® Disposable Charcoal Disks absorbs the volatile 35S radioactive component before it can escape from the Petri dish during metabolic labeling experiments. The sterile filter paper disks are saturated with activated charcoal. Prior to metabolic labeling, the disk is set in the top half of the 100mm Petri dish lid. Following the addition of the 35S - labeled compounds to the cell culture fluid, the Petri lid with the activated charcoal disk is placed over the bottom half of the Petri dish.. The volatile radioactive compounds emitted in the metabolic labeling experiment are absorbed and immobilized by the charcoal disk. Dynamic Duo! Combine the Charcoal Disks with the Charcoal Cartridges. Set the Cartridges on incubator shelves to capture volatile radiolabels that may not have been captured by Petri dishes safeguarded with Charcoal Disks.
  • Efficiently capture volatile radioactive compounds
  • Activated charcoal filter paper
  • Ideal for use in Petri dish-based labeling experiments

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service