Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B9541

SAFC

tert-Butoxy(chloro)diphenylsilane

≥97% (GC)

Synonym(s):

tert-Butoxy-diphenylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClOSi
CAS Number:
Molecular Weight:
290.86
Beilstein:
5037778
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (GC)

contains

~0.5% CaCO3 as stabilizer

density

1.076 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)O[Si](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C16H19ClOSi/c1-16(2,3)18-19(17,14-10-6-4-7-11-14)15-12-8-5-9-13-15/h4-13H,1-3H3

InChI key

MSZDOMFSWXWKTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gillard, J.W., et al.
The Journal of Organic Chemistry, 53, 2602-2602 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service