B9014
Biocytin hydrazide
≥90%
Synonym(s):
Nε-(+)-Biotinyl-L-lysine hydrazide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H30N6O3S
CAS Number:
Molecular Weight:
386.51
Beilstein:
2179522
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
≥90%
storage temp.
−20°C
SMILES string
[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](N)C(=O)NN)[C@@]1([H])NC(=O)N2
Looking for similar products? Visit Product Comparison Guide
Application
Reactive derivative for labeling proteins and glycoproteins with biotin.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service