Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B9009

Sigma-Aldrich

Bismuth nitrate oxide

Synonym(s):

Bismuth oxynitrate, Bismuth(III) nitrate basic, Bismuthyl nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi(O)NO3
CAS Number:
Molecular Weight:
286.98
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

(4BiNO3(OH)2 • BiO(OH))

SMILES string

O[Bi]=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O

InChI

1S/5Bi.4NO3.9H2O.O/c;;;;;4*2-1(3)4;;;;;;;;;;/h;;;;;;;;;9*1H2;/q+1;4*+3;4*-1;;;;;;;;;;/p-9

InChI key

OAVDTZZNYFIIGB-UHFFFAOYSA-E

Looking for similar products? Visit Product Comparison Guide

replaced by


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service