Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B8930

Sigma-Aldrich

BW373U86 dihydrobromide

powder

Synonym(s):

(±)-[1(S*),2α,5β]-4-[[2,5-Dimethyl-4-(2-propenyl)-1-piperazinyl](3-hydroxyphenyl)methyl]-N,N-diethylbenzamide dihydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37N3O2 · 2HBr
Molecular Weight:
597.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

solubility

H2O: 30 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

Br[H].Br[H].CCN(CC)C(=O)c1ccc(cc1)[C@@H](N2C[C@@H](C)N(CC=C)C[C@@H]2C)c3cccc(O)c3

InChI

1S/C27H37N3O2.2BrH/c1-6-16-29-18-21(5)30(19-20(29)4)26(24-10-9-11-25(31)17-24)22-12-14-23(15-13-22)27(32)28(7-2)8-3;;/h6,9-15,17,20-21,26,31H,1,7-8,16,18-19H2,2-5H3;2*1H/t20-,21+,26-;;/m1../s1

InChI key

ABMNQBWHHIYHEV-PKENUSFYSA-N

Biochem/physiol Actions

Potent, non-peptide δ-opioid receptor agonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Childers et al.
Molecular pharmacology, 44(4), 827-834 (1993-10-01)
BW373U86 is a potent and highly selective nonpeptidic agonist for delta-opioid receptors. To determine its ability to couple with G protein-linked second messenger systems, this study examined the effects of BW373U86 on the inhibition of adenylyl cyclase and the stimulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service