Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B8633

Sigma-Aldrich

Benzoin

Synonym(s):

α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone, 2-Hydroxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
391839
EC Number:
MDL number:
UNSPSC Code:
12352103

bp

194 °C/12 mmHg (lit.)

mp

134-138 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service