Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B8435

Sigma-Aldrich

BiCAPPA

≥97% (HPLC), solid

Synonym(s):

N,N′-Bis(6-chloro-2-methoxy-9-acridinyl)-1,4-piperazinedipropanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H40Cl2N6O2
CAS Number:
Molecular Weight:
683.67
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

solid

storage condition

under inert gas

color

yellow

solubility

DMSO: ~10 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc2nc3cc(Cl)ccc3c(NCCCN4CCN(CCCNc5c6ccc(Cl)cc6nc7ccc(OC)cc57)CC4)c2c1

InChI

1S/C38H40Cl2N6O2/c1-47-27-7-11-33-31(23-27)37(29-9-5-25(39)21-35(29)43-33)41-13-3-15-45-17-19-46(20-18-45)16-4-14-42-38-30-10-6-26(40)22-36(30)44-34-12-8-28(48-2)24-32(34)38/h5-12,21-24H,3-4,13-20H2,1-2H3,(H,41,43)(H,42,44)

InChI key

GUTKNTSGEOMFPZ-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of the formation of abnormal ß-rich isoform of prion protein PrPSc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barnaby C H May et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(6), 3416-3421 (2003-03-11)
Prion diseases are characterized by an accumulation of PrP(Sc), a misfolded isoform of the normal cellular prion protein, PrP(C). We previously reported the bioactivity of acridine-based compounds against PrP(Sc) replication in scrapie-infected neuroblastoma cells and now report the improved potency
Christopher A Ross et al.
Nature medicine, 10 Suppl, S10-S17 (2004-07-24)
Neurodegenerative diseases such as Alzheimer's disease (AD), Parkinson's disease (PD), Huntington's disease (HD), amyotrophic lateral sclerosis (ALS) and prion diseases are increasingly being realized to have common cellular and molecular mechanisms including protein aggregation and inclusion body formation. The aggregates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service