Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B7684

Sigma-Aldrich

Monoclonal Anti-Breast Cancer Resistance Protein antibody produced in mouse

250 μg/mL, clone BXP-34, tissue culture supernatant, aqueous solution

Synonym(s):

Anti-ABCG2, Anti-BCRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

BXP-34, monoclonal

form

aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

concentration

250 μg/mL

technique(s)

immunocytochemistry: 1:20-1:50 using acetone-fixed cytospin preparations
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable (not suitable)
immunohistochemistry (frozen sections): 1:20 using acetone-fixed sections

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCG2(9429)

Looking for similar products? Visit Product Comparison Guide

Specificity

Reacts with an internal epitope of BCRP. Does not react with the human MDR1, MRP1, MRP2, MRP5 gene products.

Immunogen

mitoxanthrone resistant, BCRP overexpressing cell line MCF7 MR.

Physical form

Solution in serum-free culture medium containing 1% bovine serum albumin and 0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service