Skip to Content
MilliporeSigma
All Photos(1)

Documents

B7531

Sigma-Aldrich

Restriction Enzyme Buffer SA

10 ×

Synonym(s):

Restriction Endonuclease Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

grade

for molecular biology

description

10× buffer contains 330 mM Tris-acetate, pH 7.9, 100 mM Mg acetate, 660 mM potassium acetate, 5 mM dithiothreitol at 37 °C

form

buffered aqueous solution

concentration

10 ×

storage temp.

−20°C

SMILES string

CC(C)CC(CNC(Cc1ccccc1)C(N)=O)NC(=O)C(Cc2cnc[nH]2)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc3c[nH]c4ccccc34)NC(=O)C(CCC(N)=O)NC(=O)C(N)Cc5c[nH]c6ccccc56)C(C)C

InChI

1S/C58H77N15O9/c1-32(2)21-39(29-65-46(52(61)76)22-35-13-7-6-8-14-35)69-57(81)48(25-38-28-62-31-67-38)70-50(75)30-66-58(82)51(33(3)4)73-53(77)34(5)68-56(80)47(24-37-27-64-44-18-12-10-16-41(37)44)72-55(79)45(19-20-49(60)74)71-54(78)42(59)23-36-26-63-43-17-11-9-15-40(36)43/h6-18,26-28,31-34,39,42,45-48,51,63-65H,19-25,29-30,59H2,1-5H3,(H2,60,74)(H2,61,76)(H,62,67)(H,66,82)(H,68,80)(H,69,81)(H,70,75)(H,71,78)(H,72,79)(H,73,77)

InChI key

YNZJHLFHFNCZIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service