Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B7020

Sigma-Aldrich

N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide acetate salt

factor Xa substrate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC(O)=O.CC[C@H](C)[C@H](NC(=O)c1ccccc1)C(=O)N[C@@H](CCC(O)=O)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C32H43N9O9.C2H4O2/c1-3-19(2)27(40-28(45)20-8-5-4-6-9-20)31(48)39-24(15-16-26(43)44)29(46)36-18-25(42)38-23(10-7-17-35-32(33)34)30(47)37-21-11-13-22(14-12-21)41(49)50;1-2(3)4/h4-6,8-9,11-14,19,23-24,27H,3,7,10,15-18H2,1-2H3,(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,40,45)(H,43,44)(H4,33,34,35);1H3,(H,3,4)/t19-,23-,24-,27-;/m0./s1

InChI key

ITXQNZUGLZNGBM-PJSBSAQXSA-N

Application

Factor Xa substrate

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemker, H.C.
Handbook of Synthetic Substrates, 57-57 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service