Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6934

Sigma-Aldrich

BeKm-1

lyophilized powder, >98% (HPLC), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

Assay

>98% (HPLC)

form

lyophilized powder

mol wt

apparent mol wt ~4 kDa by SDS-PAGE

solubility

aqueous buffer: soluble

storage temp.

−20°C

Amino Acid Sequence

Arg-Pro-Thr-Asp-Ile-Lys-Cys-Ser-Glu-Ser-Tyr-Gln-Cys-Phe-Pro-Val-Cys-Lys-Ser-Arg-Phe-Gly-Lys-Thr-Asn-Gly-Arg-Cys-Val-Asn-Gly-Phe-Cys-Asp-Cys-Phe s-Asp-Cys-Phe [Disulfide bridges: 7-28, 13-33, 17-35]

Biochem/physiol Actions

Specifically blocks ERG1 K+ channels.

Other Notes

Major component in Mesobuthus eupeus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service