Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6521

Sigma-Aldrich

2′-3′-O-(4-Benzoylbenzoyl)guanosine 5′-triphosphate triethylammonium salt

~95%

Synonym(s):

Benzoylbenzoyl-GTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24N5O16P3
CAS Number:
Molecular Weight:
731.39
MDL number:
UNSPSC Code:
12352204

Assay

~95%

storage temp.

−20°C

SMILES string

CCN(CC)CC.NC1=Nc2c(ncn2[C@@H]3O[C@H](COC(=O)c4ccc(c(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)c4)C(=O)c5ccccc5)[C@@H](O)[C@H]3O)C(=O)N1

Other Notes

Mixed isomers

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service