Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6312

Sigma-Aldrich

14-3-3 Beta histidine-tagged human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3 β, 14-3-3 protein Beta, GW128, HS1, KCIP-1 protein 1054, YWHAB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... YWHAB(7529)

Biochem/physiol Actions

14-3-3 proteins are a multifunctional protein family comprising seven mammalian isoforms that interact with over two hundred different intracellular molecules including kinases, phosphatases, transcription factors, scaffold proteins, and DNA. 14-3-3 proteins are involved in a multitude of biological functions and play a regulatory role in processes such as apoptotic cell death, mitogenic signal transduction, and cell cycle control. 14-3-3 β participates in various processes, among which is the positive regulation of cell cycle progression and tumorigenesis. Over-expression of 14-3-3 β in NIH-3T3 cells confers tumorigenicity in nude mice via activation of the mitogen-activated protein kinase (MAPK) cascade.
14-3-3 proteins are involved in a multitude of biological functions and play a regulatory role in processes such as apoptotic cell death, mitogenic signal transduction, and cell cycle control.

Physical form

Supplied as a 0.2 μm filtered solution in 50 mM Tris, pH 7.6, 1 mM EDTA, 1 mM DTT and 20% glycerol (w/v).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service