Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6005

Supelco

Batyl alcohol

≥98%

Synonym(s):

1-O-Octadecyl-rac-glycerol, rac-Glycerol 1-octadecyl ether, DL-3-Octadecyloxy-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
344.57
Beilstein:
1725677
EC Number:
MDL number:
UNSPSC Code:
41116107

Assay

≥98%

mp

71-73 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOCC(O)CO

InChI

1S/C21H44O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24-20-21(23)19-22/h21-23H,2-20H2,1H3

InChI key

OGBUMNBNEWYMNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service