Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B5667

Sigma-Aldrich

5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt

~97% (HPLC)

Synonym(s):

Magenta phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrClNO4P · C7H9N
CAS Number:
Molecular Weight:
433.62
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

~97% (HPLC)

Quality Level

form

powder

solubility

DMF: 50 mg/mL, clear, orange

storage temp.

−20°C

SMILES string

Cc1ccc(N)cc1.OP(O)(=O)Oc2c[nH]c3cc(Cl)c(Br)cc23

InChI

1S/C8H6BrClNO4P.C7H9N/c9-5-1-4-7(2-6(5)10)11-3-8(4)15-16(12,13)14;1-6-2-4-7(8)5-3-6/h1-3,11H,(H2,12,13,14);2-5H,8H2,1H3

InChI key

WUHVYTJTZAKPOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorota Rybaczek et al.
Histochemistry and cell biology, 135(3), 263-280 (2011-02-25)
Here, we demonstrate that in HeLa cells, Ser317 of Chk1 undergoes phosphorylation in response to replication stress induced by hydroxyurea. We also demonstrate the existence of constitutive (interphase and mitotic) Chk1 kinase phosphorylation, the translocation of its phosphorylated form from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service