Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B5435

Sigma-Aldrich

BIA 2-093

≥98% (HPLC), solid

Synonym(s):

S-(−)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$241.00

$241.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$241.00

About This Item

Empirical Formula (Hill Notation):
C17H16N2O3
CAS Number:
Molecular Weight:
296.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$241.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ~21 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CC(=O)O[C@H]1Cc2ccccc2N(C(N)=O)c3ccccc13

InChI

1S/C17H16N2O3/c1-11(20)22-16-10-12-6-2-4-8-14(12)19(17(18)21)15-9-5-3-7-13(15)16/h2-9,16H,10H2,1H3,(H2,18,21)/t16-/m0/s1

InChI key

QIALRBLEEWJACW-INIZCTEOSA-N

Biochem/physiol Actions

Blocker of voltage-gated sodium channels; significantly blocks excitatory amino acid (glutamate and aspartate) release.
Blocks voltage-gated sodium channels and excitatory amino acid release.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Bonifácio et al.
Epilepsia, 42(5), 600-608 (2001-05-31)
BIA 2-093 [(S)-(-)-10-acetoxy-10,11-dihydro-5H-dibenz/b,f/azepine-5-carboxamide] is endowed with an anticonvulsant potency similar to that of carbamazepine (CBZ), but produces less cognitive and motor impairment. This study evaluated whether voltage-gated sodium channels (VGSCs) are a primary locus for the action of BIA 2-093.
António Parada et al.
Neurochemistry international, 40(5), 435-440 (2002-02-01)
(S)-(-)-10-acetoxy-10,11-dihydro-5H-dibenz/b,f/azepine-5-carboxamide (BIA 2-093) is endowed with high anticonvulsant activity and shares with carbamazepine (CBZ) and oxcarbazepine (OXC) the capability to inhibit voltage-gated sodium channels (VGSC). The present study was aimed to compare the effects of BIA 2-093, CBZ and OXC
Sara Krarup et al.
European journal of pharmacology, 886, 173413-173413 (2020-08-08)
The antiepileptic sodium channel blocker, carbamazepine, has long been known to be able to attenuate cAMP signals. This could be of clinical importance since cAMP signaling has been shown to be involved in epileptogenesis and seizures. However, no information on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service