Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4801

Millipore

Buffered charcoal yeast extract agar base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Application

Used for the selective cultivation of Legionella.

Other Notes

Ingredients (g/L)
Yeast extract, 10.00
Charcoal activated, 2.00
ACES buffer, 10.00
α-Ketoglutarate monopotassium salt, 1.00
Agar, 17.00

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Feeley et al.
Journal of clinical microbiology, 10(4), 437-441 (1979-10-01)
Charcoal-yeast extract agar is a new bacteriological medium that supports excellent growth of the Legionella pneumophila. It results from modifications made in an existing L. pneumophila medium, F-G agar. Yeast extract, instead of an acid hydrolysate of casein, serves as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service