Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4778

Sigma-Aldrich

Boc-Gln-Gly-Arg-7-amido-4-methylcoumarin hydrochloride

~95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40N8O8 · HCl
CAS Number:
Molecular Weight:
653.13
UNSPSC Code:
12352204

Assay

~95%

storage temp.

−20°C

Substrates

Substrate for Factor XIIa

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kawabata et al.
European journal of biochemistry, 172(1), 17-25 (1988-02-15)
Seventy-four peptide amides of 7-amino-4-methylcoumarin (Mec) of the type Boc-Xaa-Yaa-Arg-NH-Mec were newly synthesized and tested to find specific substrates for blood-clotting proteases and trypsin. The Xaa and Yaa residues of these substrates have been replaced by 12 and 15 different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service