Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4022

Sigma-Aldrich

Phenacyl 4-(bromomethyl)phenylacetate

Synonym(s):

4-(Bromomethyl)phenylacetic acid phenacyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15BrO3
CAS Number:
Molecular Weight:
347.20
Beilstein:
2292032
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

84-87 °C (lit.)

SMILES string

BrCc1ccc(CC(=O)OCC(=O)c2ccccc2)cc1

InChI

1S/C17H15BrO3/c18-11-14-8-6-13(7-9-14)10-17(20)21-12-16(19)15-4-2-1-3-5-15/h1-9H,10-12H2

InChI key

BUQPIKAEKYNDAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for preparation of aminoacyloxymethyl PAM resins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitchell, A.R., et al.
The Journal of Organic Chemistry, 43, 2845-2845 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service