Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2920

Sigma-Aldrich

Benzoic acid-ring-UL-14C

aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
14C6H5COOH
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous solution

extent of labeling

1-15 mCi per mmol

mass shift

M+12

storage temp.

2-8°C

SMILES string

OC(=O)[14c]1[14cH][14cH][14cH][14cH][14cH]1

Looking for similar products? Visit Product Comparison Guide

Packaging

serum bottle

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service