Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2886

Sigma-Aldrich

(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate

Synonym(s):

BOP, BOP Reagent, Castros reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22F6N6OP2
CAS Number:
Molecular Weight:
442.28
Beilstein:
4287025
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

>130 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)[P+](On1nnc2ccccc12)(N(C)C)N(C)C

InChI

1S/C12H22N6OP.F6P/c1-15(2)20(16(3)4,17(5)6)19-18-12-10-8-7-9-11(12)13-14-18;1-7(2,3,4,5)6/h7-10H,1-6H3;/q+1;-1

InChI key

MGEVGECQZUIPSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Activating reagent for synthesis of peptides and amino acid derivatives

replaced by

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Castro, B, et al.
Tetrahedron Letters, 1219, 1219-1219 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service