Skip to Content
MilliporeSigma
All Photos(1)

Documents

B2625

Sigma-Aldrich

Benzaldehyde p-nitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O2
CAS Number:
Molecular Weight:
241.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(N\N=C\c2ccccc2)cc1

InChI

1S/C13H11N3O2/c17-16(18)13-8-6-12(7-9-13)15-14-10-11-4-2-1-3-5-11/h1-10,15H/b14-10+

InChI key

NOIFWEYOLLHIMW-GXDHUFHOSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service