Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2545

Sigma-Aldrich

Benzoic acid-carboxy-14C

aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H514COOH
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous solution

extent of labeling

1-20 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

O[14C](=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)/i7+2

InChI key

WPYMKLBDIGXBTP-WGGUOBTBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

serum bottle

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service