Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2026

Sigma-Aldrich

des-Pro2-Bradykinin

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H66N14O10
CAS Number:
Molecular Weight:
963.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)NCC(=O)NC(Cc2ccccc2)C(=O)NC(CO)C(=O)N3CCCC3C(=O)NC(Cc4ccccc4)C(=O)NC(CCCNC(N)=N)C(O)=O

Gene Information

mouse ... Tac1(21333)

Amino Acid Sequence

Arg-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Biochem/physiol Actions

Potentiates activity on bradykinin-induced contraction of guinea pig ileum and hypertension in rats

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiating effects of bradykinin-related substances on bradykinin-induced contraction of guinea pig ileum and hypotension in rats.
M Naruse et al.
Chemical & pharmaceutical bulletin, 29(11), 3369-3374 (1981-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service