Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B1011

Sigma-Aldrich

Boc-Glu-Glu-Leu methyl ester

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H37N3O10
CAS Number:
Molecular Weight:
503.54
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Boc-Glu-Glu-Leu methyl ester, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

COC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)OC(C)(C)C

InChI

1S/C22H37N3O10/c1-12(2)11-15(20(32)34-6)24-18(30)13(7-9-16(26)27)23-19(31)14(8-10-17(28)29)25-21(33)35-22(3,4)5/h12-15H,7-11H2,1-6H3,(H,23,31)(H,24,30)(H,25,33)(H,26,27)(H,28,29)/t13-,14-,15-/m0/s1

InChI key

QJKPYQOKXLMHNY-KKUMJFAQSA-N

Biochem/physiol Actions

Boc-Glu-Glu-Leu methyl ester (Boc-Glu-Glu-Leu-OMe) is an N-terminal and C-terminal blocked derivative of the tripeptide L-Glu-LGlu-L-Leu.

Substrates

Possible substrate for Vitamin K-dependent carboxylation studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Pottorf et al.
Journal of medicinal chemistry, 30(2), 445-448 (1987-02-01)
The synthesis of the amino acid sequence found in bovine prothrombin precursor 13-29 (PTP 13-29) has been achieved by solid-phase synthesis of the bis(acetamidomethyl)-protected linear peptide followed by cyclization to the monomeric disulfide. Synthesis of the disulfide bond was achieved
C Vermeer et al.
Haematologia, 18(2), 71-97 (1985-01-01)
Vitamin K-dependent carboxylase is found in the liver, where it is involved in the synthesis of four blood coagulation factors and protein C. The hepatic enzyme has partly been purified and several mechanisms have been postulated for the vitamin K-dependent
Vitamin K-dependent carboxylation.
P A Friedman et al.
The International journal of biochemistry, 19(1), 1-7 (1987-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service