AXYVT250RLAB
Pipette tips for Velocity, robotic
size 250 μL, pkg of 5x960tips/cs, racked in bottom sealed rack
Synonym(s):
Pipette tips for Velocity
Select a Size
$445.00
Select a Size
About This Item
Corning
$445.00
Recommended Products
sterility
non-sterile
manufacturer/tradename
Axygen Inc VT-250-R-LAB
Corning
packaging
pkg of 5x960tips/cs
size
250 μL
volume
250 μL
Features and Benefits
- For greater consistency and performance they are assembled via automation
- Tips are RNAse/DNAse free
- Tips are made of polypropylene and are non-pyrogenic
- Compatible with Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST and 16ST head
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)