Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYTR333C

Corning® Axygen® Axypet® Universal Fit Pipet Tips

size 250 μL, pkg of 20x1000tips/cs, clear

Synonym(s):

Pipette reference tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
volume:
250 μL
packaging:
pkg of 20x1000tips/cs
manufacturer/tradename:
Axygen Inc TR-333-C
Corning

manufacturer/tradename

Axygen Inc TR-333-C
Corning

packaging

pkg of 20x1000tips/cs

size

250 μL

volume

250 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Legal Information

AxyPet is a registered trademark of Axygen, Inc.
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service