Key Documents
AXYTGL200R
Ultra micro gel loader pipette tips
diam. 0.66 mm, capacity 200 μL
Synonym(s):
pipet tips
About This Item
UNSPSC Code:
41121600
volume:
200 μL
sterility:
non-sterile
feature:
gel loader tips
packaging:
case of 480 tips racked
manufacturer/tradename:
Axygen Inc
Corning
sterility
non-sterile
feature
gel loader tips
packaging
case of 480 tips racked
manufacturer/tradename
Axygen Inc
Corning
capacity
200 μL
diam.
0.66 mm
volume
200 μL
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)