Skip to Content
MilliporeSigma
All Photos(1)

Documents

AXYTF300

Pipette filter tips

for P-2, ultramicro, size 0.5-10 μL, pkg of 10x1000tips/cs

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-300
Corning

packaging

pkg of 10x1000tips/cs

size

0.5-10 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® Microvolume - Gilson-Style P2/P10 filter tips are optimized for use in sensitive biological applications for accurate results.
  • Features an aerosol-resistant filter to prevent the possible cross-contamination of samples.
  • The polyethylene aerosol filter is inserted robotically into the pipette tip to create a physical barrier in the tip for protecting samples from nuclease contamination.

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service