AXYT5000C
Pipette tips
for Gilson® pipettors, size 5 mL, pkg of 10x250tips/cs
Synonym(s):
Disposable Pipette Tips, Laboratory Pipette Tips, Pipetting Tips
Select a Size
$454.00
Select a Size
About This Item
RNase free
barrier: no
endotoxin free
filter: no
Corning
$454.00
Recommended Products
material
clear
sterility
non-sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc T-5000-C
Corning
packaging
pkg of 10x250tips/cs
size
5 mL
volume
5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service