Skip to Content
MilliporeSigma
All Photos(1)

Documents

AXYT300RS

Pipette tips, for P2/P10

racked, size 0.5-10 μL, clear, pkg of 5x960tips/cs, sterile

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc T-300-R-S
Corning

packaging

pkg of 5x960tips/cs

size

0.5-10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Axygen® Universal Tips provide the following benefits:
  • Tips fit a broad range of single and multi-channel pipettors
  • Compatible with most of the well-known brands of pipettors
  • Tips are made of polypropylene and are non-pyrogenic
  • Tips are RNAse and DNAse free
  • Flexible walls and a series of internal sealing rings provides good fitting which requires less force to load and eject
  • Microvolume
  • Low-risk of sample loss due to high fluid retention
  • Low-risk of cross-contamination
  • Higher yield, accuracy, and precision

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service