Key Documents
AXYSCT200RS
Screw cap tubes and caps with O-rings, conical bottom
capacity 2.0 mL, red, sterile
Synonym(s):
centrifuge tubes
About This Item
UNSPSC Code:
41121800
material
red
screw top cap
sterility
sterile
feature
RCF 20,000 × g
packaging
pack of 500 tubes
case of 8 packs
manufacturer/tradename
Axygen Inc
Corning
capacity
2.0 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)