Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYSCT150CS

Screw cap tube

size 1.5 mL, clear, pkg of 8x500caps/cs, sterile

Synonym(s):

Tube with Screw Cap

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$1,170.00

$1,170.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$1,170.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

$1,170.00


Please contact Customer Service for Availability

material

conical bottom tube
screw top cap

sterility

sterile

manufacturer/tradename

Axygen Inc SCT-150-C-S
Corning

packaging

pkg of 8x500caps/cs

parameter

20000 × g max. RCF

size

1.5 mL

color

clear
clear

fitting

cap (screw cap)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® microcentrifuge tube features a screw cap for an efficient seal.
  • The screw cap O-rings protects sample from spilling, cross-contamination and evaporation.
  • The tubes are free of RNase-/DNase and nonpyrogenic.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service