Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYR80R

Corning® Axygen® 80 Well Microtube Rack

size 80 wells, for 1.5/2.0 mL MicroTube, red, 25 ea of

Synonym(s):

Microtube rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$151.00

$151.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$151.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.22

$151.00


Please contact Customer Service for Availability

manufacturer/tradename

Axygen Inc R-80-R
Corning

packaging

25 ea of

size

80 wells

color

red

Looking for similar products? Visit Product Comparison Guide

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service