AXYPCR96M2HSR
PCR amplification plate microplate
half skirt with a single notch, size 96 wells, red, pkg of 5x10plates/cs
Synonym(s):
96 multiwell plate
Select a Size
$410.00
Select a Size
About This Item
$410.00
Recommended Products
material
conical bottom wells
feature
skirt
manufacturer/tradename
Axygen Inc PCR-96M2-HS-R
Corning
packaging
pkg of 5x10plates/cs
size
96 wells
color
red
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service